Skip to Content
Merck
All Photos(1)

Key Documents

183482

Sigma-Aldrich

β-Ethylphenethyl alcohol

98%

Synonym(s):

2-Phenyl-1-butanol

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
C6H5CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

234-236 °C (lit.)

density

0.959 g/mL at 25 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

CCC(CO)c1ccccc1

InChI

1S/C10H14O/c1-2-9(8-11)10-6-4-3-5-7-10/h3-7,9,11H,2,8H2,1H3

InChI key

DNHNBMQCHKKDNI-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

215.6 °F - closed cup

flash_point_c

102.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Mezzetti et al.
Chemistry & biology, 12(4), 427-437 (2005-04-27)
Synthetic chemists often exploit the high enantioselectivity of lipases to prepare pure enantiomers of primary alcohols, but the molecular basis for this enantioselectivity is unknown. The crystal structures of two phosphonate transition-state analogs bound to Burkholderia cepacia lipase reveal this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service