Skip to Content
MilliporeSigma
All Photos(1)

Documents

136417

Sigma-Aldrich

4-(Methylsulfonyl)benzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2C6H4CO2H
CAS Number:
Molecular Weight:
200.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

268-271 °C (lit.)

SMILES string

CS(=O)(=O)c1ccc(cc1)C(O)=O

InChI

1S/C8H8O4S/c1-13(11,12)7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

AJBWNNKDUMXZLM-UHFFFAOYSA-N

General description

4-(Methylsulfonyl)benzoic acid has pKa value of 3.48 and 8.36 in water and methanol at 25°C.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dissociation constants of neutral and charged acids in methyl alcohol. The acid strength resolution.
Rived F, et al.
Analytica Chimica Acta, 374(2), 309-324 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service