Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

117994

Sigma-Aldrich

2,4-Dinitrodiphenylamine

98%

Synonym(s):

2,4-Dinitro-N-phenylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3NHC6H5
CAS Number:
Molecular Weight:
259.22
Beilstein/REAXYS Number:
8588595
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

159-161 °C (lit.)

solubility

acetone: soluble 25 mg/mL, clear, orange to red

SMILES string

[O-][N+](=O)c1ccc(Nc2ccccc2)c(c1)[N+]([O-])=O

InChI

1S/C12H9N3O4/c16-14(17)10-6-7-11(12(8-10)15(18)19)13-9-4-2-1-3-5-9/h1-8,13H

InChI key

RHTVQEPJVKUMPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dinitrodiphenylamine forms an efficient anionic chromogenic chemosensor[1]. It was anaerobically metabolized in sediment-water batch enrichments inoculated with mud from the German North Sea coast[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanderléia Gava Marini et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(2), 799-806 (2010-01-01)
Solutions of 2,4-dinitrodiphenylamine (1) in dimethylsulfoxide (DMSO) are colorless but upon deprotonation they become red. Addition of various anionic species (HSO(4)(-), H(2)PO(4)(-), NO(3)(-), CN(-), CH(3)COO(-), F(-), Cl(-), Br(-), and I(-)) to solutions of 1 revealed that only CN(-), F(-), CH(3)COO(-)
O Drzyzga et al.
Applied and environmental microbiology, 61(9), 3282-3287 (1995-09-01)
2-Nitrodiphenylamine, 4-nitrodiphenylamine, and 2,4-dinitrodiphenylamine were anaerobically metabolized in sediment-water batch enrichments inoculated with mud from the German North Sea coast. The first intermediate in 2,4-dinitrodiphenylamine degradation was 2-amino-4-nitrodiphenylamine, which appeared in large (nearly stoichiometric) amounts before being completely reduced to
Djaafar Siddiq Karahacane et al.
Forensic science international, 301, 129-136 (2019-06-04)
The detection and analysis of gunshot residues expelled during the discharge of the weapon play a potential role in criminal investigations. In this work, organic gunshot residues produced by using automatic rifle Kalashnikov caliber: 7.62×39mm and semi-automatic pistol Makarov caliber:
Kristen L Reese et al.
Forensic science international, 272, 16-27 (2017-01-16)
This work demonstrates a non-targeted mass spectrometry approach for identification of organic compounds in smokeless powders. Unburned powders were removed from various commercial ammunitions of different brand, primer composition, caliber, and age. The unburned powders and corresponding fired residues were
Zuriñe Abrego et al.
The Analyst, 139(23), 6232-6241 (2014-10-11)
A method based on scanning laser ablation and inductively coupled plasma-mass spectrometry (SLA-ICPMS) and Raman micro-spectroscopy for the detection and identification of compounds consistent with gunshot residue particles (GSR) has been developed. The method has been applied to the characterization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service