Developed in collaboration with the Protein Quantitation Consortium, our AQUA™ Peptides enable accurate, efficient mass spectrometric quantitation of protein biomarkers.
- Focus your analysis on significant protein biomarkers
- Accurately quantitate low abundance proteins
- Eliminate costly, time consuming stable isotope labeling steps
- Measure site-specific phosphorylation states
- Validate gene silencing at the protein level
- Learn more on the AQUA™ Strategy and Applications
This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School.1 Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.
We offer high-purity custom peptides. Our custom peptides are stringently tested to ensure high purity (HPLC), accurate molecular weight (ESI-MS), and specific peptide content. Custom peptides are available without stable isotope labeling or with stable isotope labeling.
Designing the Optimal Sequence
Selecting an optimal AQUA™ peptide is key to your analytical success. Because your time and resources are valuable, we suggest a careful evaluation of candidate peptides. The following guidelines provide a good starting point.
Using the known specificity of trypsin and the sequence of your protein of interest, generate a complete list of its tryptic peptides. Using the guidelines below, eliminate unsuitable peptides and select peptides with optimal characteristics for AQUA™ experimentation
- Choose a peptide that is unique to your protein of interest
- Choose a tryptic peptide that resolves well by HPLC
- Avoid peptides that are too hydrophilic or too hydrophobic
- Choose a peptide that ionizes well in your MS system
- Avoid chemically reactive residues such as Tryptophan, Methionine, Cysteine, or chemically unstable sequences such as Asp-Gly, N-term Gln, N-term Asn
- Choose an amino acid from the available list to be isotopically labeled
- Peptide sequence length should be limited to 15 amino acids or less
Isotopically Labeled Amino Acids | |
---|---|
Fully Labeled,13C, 15N > 98 atom% | Mass Difference between Native Peptide and AQUA™ Peptide |
L-Arginine-13C6,15N4 | 10 Daltons |
L-Isoleucine-13C6,15N | 7 Daltons |
L-Leucine-13C6,15N | 7 Daltons |
L-Lysine-13C6,15N2 | 8 Daltons |
L-Phenylalanine-13C9,15N | 10 Daltons |
L-Proline-13C5,15N | 6 Daltons |
L-Valine-13C5,15N | 6 Daltons |
AQUA™ Peptide Specifications | |
---|---|
Amount | 5 x 1 nmol (quantification by AAA) |
Length | 5 - 30 amino acids |
Purity | >95% by RP-HPLC |
Modifications | Phosphorylation (Ser, Thr, Tyr), carboxymethylated Cys, |
Stable Isotope | Fully labeled 98 atom% 13C and 98 atom% 15N enriched amino acids, |
Quality Control | All peptides by HPLC and MS; additional analysis available upon request |
Format | Supplied lyophilized |
Highlights