Merck
  • Thermal solvent-free synthesis of chromonyl chalcones, pyrazolines and their in vitro antibacterial, antifungal activities.
Thermal solvent-free synthesis of chromonyl chalcones, pyrazolines and their in vitro antibacterial, antifungal activities.
Journal of enzyme inhibition and medicinal chemistry (2011-05-27)
Zeba N Siddiqui, Shagufta Praveen, T N Mohammed Musthafa, Anis Ahmad, Asad U Khan
ABSTRACT
A facile and ecofriendly synthesis of new chromonyl chalcones 3a-b from 3-formylchromone 1 and active methyl compounds 2a-b is reported under thermal solvent-free heating condition in good yields. The chromonyl chalcones 3a-b were used as intermediates under green condition for the synthesis of new bioactive pyrazoline derivatives 4a-f. The compounds were tested for antimicrobial activity by disk diffusion assay with slight modifications against Gram-positive, Gram-negative strains of bacteria as well as fungal strains. The investigation of antimicrobial screening revealed that compounds 3a-b and 4a-f showed antibacterial and antifungal activities.
Sigma-Aldrich
3-Formylchromone, 97%