Merck
  • Single-stranded DNA-binding proteins (SSBs) -- sources and applications in molecular biology.
Single-stranded DNA-binding proteins (SSBs) -- sources and applications in molecular biology.
Acta biochimica Polonica (2005-08-06)
Józef Kur, Marcin Olszewski, Anna Długołecka, Paweł Filipkowski
ABSTRACT
Single-stranded DNA-binding proteins (SSBs) play essential roles in DNA replication, recombination, and repair in bacteria, archaea and eukarya. The SSBs share a common core ssDNA-binding domain with a conserved OB (oligonucleotide/oligosaccharide binding) fold. This ssDNA-binding domain was presumably present in the common ancestor to all three major branches of life. In recent years, there has been an increasing interest in SSBs because they are useful for molecular biology methods and for analytical purposes. In this review, we concentrate on recent advances in the discovery of new sources of SSBs as well as certain aspects of their applications in analytical sciences.
Sigma-Aldrich
Polythymidylic acid sodium salt, homopolymer