Skip to Content
Merck
All Photos(2)

Key Documents

AV31263

Sigma-Aldrich

Anti-NFYA antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Nuclear transcription factor Y, α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

human, mouse, horse, bovine, rat, rabbit, guinea pig, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFYA(4800)

General description

NFYA is a nuclear transcription factor that forms a regulatory subunit of the trimeric complex of NF-Y that associates with CCAAT motifs in cell cycle progression genes. Hence, this gene has been implicated in cell proliferation. Furthermore, NFYA can also facilitate the self-renewal of hematopoietic stem cells (HSCs).
Rabbit Anti-NFYA antibody recognizes canine, chicken, human, mouse, rat, and bovine NFYA.

Immunogen

Synthetic peptide directed towards the C terminal region of human NFYA

Application

Rabbit Anti-NFYA antibody can be used for immunohistochemistry (4-8μg/ml using paraffin-embedded tissues) and western blot (5-8μg/ml) applications.

Biochem/physiol Actions

NFYA is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms.

Sequence

Synthetic peptide located within the following region: YLHESRHRHAMARKRGEGGRFFSPKEKDSPHMQDPNQADEEAMTQIIRVS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabella Manni et al.
Molecular biology of the cell, 19(12), 5203-5213 (2008-09-26)
NF-Y binds to CCAAT motifs in the promoter region of a variety of genes involved in cell cycle progression. The NF-Y complex comprises three subunits, NF-YA, -YB, and -YC, all required for DNA binding. Expression of NF-YA fluctuates during the
Jiang Zhu et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(33), 11728-11733 (2005-08-06)
Hematopoietic stem cell (HSC) self-renewal and differentiation are influenced through multiple pathways, including homeobox transcription factors, signaling through beta-catenin and Notch-1, telomerase, and p27. How these multiple pathways interact and are orchestrated is currently unknown. We now report that NF-Ya

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service