Skip to Content
Merck
All Photos(1)

Key Documents

XFUF07601

Millipore

Solvent-resistant Stirred Cell, for 76 mm membranes

Protein Desalting, Protein Purification, Protein Concentration

Synonym(s):

76 mm Stirred Cell, Solvent-Resistant Cell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031690
NACRES:
NB.24

material

316L stainless steel plate (base)
316L stainless steel top(s)
Nylon tube (pressure)
PTFE stir bar(s)
borosilicate glass cylinder
fluoroelastomer O-ring
high-density polyethylene funnel(s)
silicone tube (for filtrate)

Quality Level

feature

holdup volume 1 mL

parameter

10-300 mL process volume
6.2 bar max. inlet pressure (90 psi)

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

H

16.5 cm (6.5 in.)

diam.

11 cm

filter diam.

76 mm

filtration area

40 cm2

shipped in

ambient

General description

For applications requiring compatibility with solvents, this Millipore stirred cell is available in two sizes. Certain aldehydes, ketones (e. g., acetone) and aliphatic ethers/esters may reduce the life of the fluorocarbon O-rings. Please contact Millipore for more information.

  • Borosilicate glass cylinder and PTFE components for broad compatibility
  • Autoclavable with membrane in-place
  • Top plate opening provides access to contents without dismantling
  • Few components, so it′s easy to clean and assemble
  • For 76 mm disc filters


Applications

Concentration and Buffer Exchange Involving Organic Solvents

Application

Protein Desalting, Protein Purification, Protein Concentration

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoping Wang et al.
Chemosphere, 256, 127099-127099 (2020-05-30)
Membrane fouling is an issue of concern due to the hydrophobic properties of polyethersulfone (PES) membrane when applied in water treatment. In this work, a facile hydrothermal method was utilized to synthesize hierarchical flower-like structured molybdenum disulfide nanosheets (HF-MoS2 NSs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service