Merck
  • Increased expression of TNFRSF14 indicates good prognosis and inhibits bladder cancer proliferation by promoting apoptosis.
Increased expression of TNFRSF14 indicates good prognosis and inhibits bladder cancer proliferation by promoting apoptosis.
Molecular medicine reports (2018-08-02)
Yu-Di Zhu, Ming-Yue Lu
ABSTRACT
Despite advances in management, bladder cancer remains a principal cause of cancer‑associated complications. Tumor necrosis factor receptor superfamily member 14 (TNFRSF14) is dysregulated in certain types of cancer; however, limited data are available on the expression and function of TNFRSF14 in bladder cancer. In the present study, the aim was to evaluate the expression and biological functions of TNFRSF14 in bladder cancer. Firstly, the expression levels of TNFRSF14 in bladder cancer tissue were examined using The Cancer Genome Atlas (TCGA) database. Secondly, reverse transcription‑quantitative polymerase chain reaction was utilized to investigate the expression levels of TNFRSF14 in the T24, SW780 and EJ‑M3 bladder cancer cell lines. Transfection and Cell Counting kit‑8 (CCK‑8) assay was used to evaluate whether TNFRSF14 overexpression or silencing would have an effect on cell proliferation of T24 and EJ‑M3 cells. In addition, TNFRSF14‑induced apoptotic cells were identified using Annexin V‑fluorescein isothiocyanate and propidium iodide staining. Western blot analysis was used to detect proteins associated with the phosphatidylinositol 3‑kinase pathway. According to the TCGA dataset, the expression levels TNFRSF14 were decreased in bladder cancer tissue compared with in normal control samples. Patients with bladder cancer exhibiting low expression levels of TNFRSF14 had a worse prognosis compared to those with high expression levels of TNFRSF14. Overexpression of TNFRSF14 in T24 cells led to increased apoptosis and inhibited cell proliferation in vitro. Western blotting demonstrated that TNFRSF14 overexpression increased the expression levels of caspase3‑p17 in T24 cells, but significantly decreased the expression levels of phosphorylated (p)‑protein kinase B (AKT) and P70 S6 kinase (P70). TNFRSF14 silencing in EJ‑M3 cells enhanced cell growth, inhibited cell apoptosis, increased the expression levels of p‑AKT and P70, and decreased the expression levels of caspase3‑p17. In conclusion, TNFRSF14 may serve a tumor suppressive role in bladder cancer by inducing apoptosis and suppressing proliferation, and act as a novel prognostic biomarker for bladder cancer.
Sigma-Aldrich
Anti-CASP3 (p17, Cleaved-Asp175) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p70 S6 Kinase antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody