MT51341000
Mettler Toledo DX series GSE half-cells electrode
GSE Ammonia
All Photos(1)
About This Item
Recommended Products
material
shaft (PPS)
manufacturer/tradename
Mettler-Toledo 51341000
measuring range
0.5 ... 5.0-3
temperature measuring range
0-50 °C
pH
>11
General description
Modular and versatile the METTLER TOLEDO DX series ion-selective half-cells consist of two elements: a universal shaft and an ion-specific membrane module. This module may be exchanged allowing you to measure the ion of your choice. Just order the membrane kit specific for that ion, mount the new module onto the shaft of your GSE half-cell, and you have a new GSE!
Some interference may be seen with volatile bases. ISA solution: 1 mol/L KNO3
Some interference may be seen with volatile bases. ISA solution: 1 mol/L KNO3
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)