Skip to Content
Merck

PF126

Bcl-2, C-Terminal Truncated, His•Tag®, Human, Recombinant, E. coli

Recombinant, human Bcl-2 without the 17 amino acid mitochondrial targeting sequence at the C-terminus, fused to a His•Tag sequence, and expressed in E. coli.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.77
UNSPSC Code:
12352202
Form:
liquid
Assay:
≥95% (SDS-PAGE)
Biological source:
human
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Neutralization (see biological activity))

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human Bcl-2 without the 17 amino acid mitochondrial targeting sequence at the C-terminus, fused to a His•Tag sequence, and expressed in E. coli. The truncated protein retains the ability to neutralize pro-apoptotic Bcl-2 family members.

Application

Neutralization (see biological activity)

Biochem/physiol Actions

EC₅₀ = 100-300 nM based on its ability to inhibit 54 nM caspase-8-cleaved human Bid-mediated release of cytochrome c from isolated mouse liver mitochondria

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 100 mM KCl, 10 mM HEPES, 20% glycerol, pH 8.0.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library