386023
Anti-Hsp47 Mouse mAb (M16.10A1)
liquid, clone M16.10A1, Calbiochem®
Synonym(s):
Anti-Colligin
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
M16.10A1, monoclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
mouse, porcine, chicken, canine, human, hamster, guinea pig, bovine, rat, monkey, sheep
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
isotype
IgG2b
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... SERPINH1(871)
General description
Immunogen
Application
Enzyme Immunoassay (see comments)
Immunoblotting (1 µg/ml, colorimetric)
Immunocytochemistry (1 µg/ml)
Immunofluorescence (5 µg/ml)
Immunoprecipitation (5 µg/ml)
Paraffin Sections (see comments)
Packaging
Warning
Physical form
Reconstitution
Analysis Note
3T3 cells, Human breast cancer tissue
Other Notes
Nandan, D., et al. 1998. Exp. Cell Res.179, 289.
Clark, E.P., et al. 1991. J. Biol. Chem.266, 17230.
Vaillancourt, J.P., and Cates, G.A. 1991. Biochem. J.274, 793.
Cates, G.A., et al. 1987. Biochem. Cell Biol. 65, 767.
Saga, S., et al. 1987. J. Cell Biol.105, 517.
Nagata, K., et al. 1986. J. Biol. Chem. 261, 7531.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service