Skip to Content
Merck
All Photos(1)

Key Documents

1.14414

Supelco

Copper Test Kit, colorimetric

0.05-0.5 mg/L (Cu), MQuant®

Synonym(s):

Copper Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Copper Test, colorimetric, 0.05-0.5 mg/L (Cu), for use with MCOLORTEST®

specific analyte(s)

copper

Quality Level

measuring range

0.05-0.5 mg/L (Cu)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with long test tubes 0.05 - 0.08 - 0.12 - 0.16 - 0.20 - 0.25 - 0.3 - 0.4 - 0.5 mg/l Cu MQuant®
The MQuant® Copper test (formally MColortest®) measures copper(II) ions in
  • Groundwater, surface water, and seawater
  • Drinking water
  • Wastewater and percolating water
  • Boiler and boiler feed water, cooling water
  • Electroplating wastewater
  • Food and beverages after appropriate sample pretreatment
  • Swimming-pool water

Method
In an ammoniacal medium copper(II) ions react with cuprizone to form a blue complex. The copper concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. By the addition of hydrogen peroxide copper(I) ions can be measured at the same time.
The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Using the refill pack 1.18459 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service