Skip to Content
Merck
All Photos(1)

Key Documents

Z103292

Sigma-Aldrich

Storage bottles without joints

capacity 250 mL, PTFE stopcock

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

PTFE stopcock
colorless bottle
glass bottle
round bottle

packaging

pack of 1 ea

manufacturer/tradename

Sigma-Aldrich

capacity

250 mL

stopcock size

2 mm

Looking for similar products? Visit Product Comparison Guide

General description

Clear glass, with stopcock-equipped septum-inlet.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service