跳轉至內容
Merck
  • Ibuprofen and lipoic acid conjugate neuroprotective activity is mediated by Ngb/Akt intracellular signaling pathway in Alzheimer's disease rat model.
Ibuprofen and lipoic acid conjugate neuroprotective activity is mediated by Ngb/Akt intracellular signaling pathway in Alzheimer's disease rat model.
Gerontology (2013-02-23)
Susi Zara, Marianna De Colli, Monica Rapino, Stephanie Pacella, Cinzia Nasuti, Piera Sozio, Antonio Di Stefano, Amelia Cataldi
摘要
Alzheimer's disease (AD) is a frequent form of senile dementia. Neuroglobin (Ngb) has a neuroprotective role and decreases Aβ peptide levels. Ngb, promoting Akt phosphorylation, activates cell survival involving cyclic-nucleotide response element-binding protein (CREB). A new molecule (IBU-LA) was synthetized and administered to an AD rat model to counteract AD progression. The aim of this study was to investigate the IBU-LA-mediated induction of Ngb neuroprotective and antiapoptotic activities. Brain morphology was analyzed through Bielschowsky staining, Aβ(1-40) and Ngb expression by immunohistochemistry. Akt, p-Akt, CREB and p-CREB expression was evaluated by Western blot, apoptosis through cytochrome C/Apaf 1 immunocomplex formation, and TUNEL analysis. Bielschowsky staining and Aβ(1-40) expression show few nerve connections and Aβ(1-40) expression in an Aβ sample, preserved neuronal cells and Aβ(1-40) expression lowering in an IBU sample, mostly in IBU-LA. The Ngb level decreases in Aβ samples, compared to control and IBU-LA samples. p-Akt/Akt and p-CREB/CREB ratios reveal a reduction in Aβ sample, going back to the basal level in control and IBU-LA samples. Cytochrome C/Apaf 1 co-immunoprecipitate occurs and TUNEL-positive nuclei percentage decreases in Aβ sample. Probe test performance shows an increased spatial reference memory in the IBU-LA compared to the Aβ sample; no significant differences were seen between the IBU-LA and IBU samples. This evidence reveals that IBU-LA administration has the capability to maintain a high Ngb level allowing Ngb to perform a neuroprotective and antiapoptotic role, representing a valid tool in the therapeutic strategy of AD progression.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
Anti-Neuroglobin, ~1.2 mg/mL, affinity isolated antibody, buffered aqueous solution