跳轉至內容
Merck
  • PARP Inhibitor Activity Correlates with SLFN11 Expression and Demonstrates Synergy with Temozolomide in Small Cell Lung Cancer.
PARP Inhibitor Activity Correlates with SLFN11 Expression and Demonstrates Synergy with Temozolomide in Small Cell Lung Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-07-22)
Benjamin H Lok, Eric E Gardner, Valentina E Schneeberger, Andy Ni, Patrice Desmeules, Natasha Rekhtman, Elisa de Stanchina, Beverly A Teicher, Nadeem Riaz, Simon N Powell, John T Poirier, Charles M Rudin
摘要
PARP inhibitors (PARPi) are a novel class of small molecule therapeutics for small cell lung cancer (SCLC). Identification of predictors of response would advance our understanding, and guide clinical application, of this therapeutic strategy. Efficacy of PARP inhibitors olaparib, rucaparib, and veliparib, as well as etoposide and cisplatin in SCLC cell lines, and gene expression correlates, was analyzed using public datasets. HRD genomic scar scores were calculated from Affymetrix SNP 6.0 arrays. In vitro talazoparib efficacy was measured by cell viability assays. For functional studies, CRISPR/Cas9 and shRNA were used for genomic editing and transcript knockdown, respectively. Protein levels were assessed by immunoblotting and immunohistochemistry (IHC). Quantitative synergy of talazoparib and temozolomide was determined in vitro In vivo efficacy of talazoparib, temozolomide, and the combination was assessed in patient-derived xenograft (PDX) models. We identified SLFN11, but not HRD genomic scars, as a consistent correlate of response to all three PARPi assessed, with loss of SLFN11 conferring resistance to PARPi. We confirmed these findings in vivo across multiple PDX and defined IHC staining for SLFN11 as a predictor of talazoparib response. As temozolomide has activity in SCLC, we investigated combination therapy with talazoparib and found marked synergy in vitro and efficacy in vivo, which did not solely depend on SLFN11 or MGMT status. SLFN11 is a relevant predictive biomarker of sensitivity to PARP inhibitor monotherapy in SCLC and we identify combinatorial therapy with TMZ as a particularly promising therapeutic strategy that warrants further clinical investigation. Clin Cancer Res; 23(2); 523-35. ©2016 AACR.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
抗 SLFN11 兔抗, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution