跳轉至內容
Merck
  • Reversible association of thermoresponsive gold nanoparticles: polyelectrolyte effect on the lower critical solution temperature of poly(vinyl methyl ether).
Reversible association of thermoresponsive gold nanoparticles: polyelectrolyte effect on the lower critical solution temperature of poly(vinyl methyl ether).
The journal of physical chemistry. B (2006-03-31)
Rama Ranjan Bhattacharjee, Mukut Chakraborty, Tarun K Mandal
摘要
Thermoresponsive gold nanoparticles (GNPs) have been prepared by the borohydride reduction of gold salt in the presence of water-soluble polymer, poly(vinyl methyl ether) (PVME). The PVME-coated GNPs (PVME-GNPs) have been assembled into large aggregates in the presence of polyelectrolytes, viz., poly(sodium-4 styrene sulfonate) and sodium salt of carboxymethylcellulose at low pH by raising the solution temperature from 20 to 40 degrees C. Increase of temperature triggers the interparticle association due to hydrophobic interaction of pendent methyl group of the surface adsorbed PVME. This assembly process is reversible with respect to temperature and pH of the medium and was studied by monitoring the change in surface plasmon resonance band of PVME-GNPs. Three-dimensional assemblies of various architectures, depending on the concentration of polyelectrolytes, were observed through transmission electron microscopy. A mechanistic model has been suggested for the reversible assembly formation that suits well with the experimental observations. The changes in optical properties of the PVME-GNPs due to their aggregation/disaggregation enabled us to use it as an effective tool to monitor the change in lower critical solution temperature (LCST) of PVME in the presence of polyelectrolytes due to interpolymer complexation at low pH. This result agrees well with the variation of LCST of pure aqueous PVME solution in the presence of polyelectrolytes measured by conventional turbidimetric technique.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
氯化金 (III) 三水合物, ≥99.9% trace metals basis