跳轉至內容
Merck
  • Ideal probe single-molecule experiments reveal the intrinsic dynamic heterogeneity of a supercooled liquid.
Ideal probe single-molecule experiments reveal the intrinsic dynamic heterogeneity of a supercooled liquid.
Proceedings of the National Academy of Sciences of the United States of America (2015-04-01)
Keewook Paeng, Heungman Park, Dat Tien Hoang, Laura J Kaufman
摘要
The concept of dynamic heterogeneity and the picture of the supercooled liquid as a mosaic of environments with distinct dynamics that interchange in time have been invoked to explain the nonexponential relaxations measured in these systems. The spatial extent and temporal persistence of these regions of distinct dynamics have remained challenging to identify. Here, single-molecule fluorescence measurements using a probe similar in size and mobility to the host o-terphenyl unambiguously reveal exponential relaxations distributed in time and space and directly demonstrate ergodicity of the system down to the glass transition temperature. In the temperature range probed, at least 200 times the structural relaxation time of the host is required to recover ensemble-averaged relaxation at every spatial region in the system.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
邻二苯基苯, 99%