跳轉至內容
Merck
  • miR-338-3p suppresses gastric cancer progression through a PTEN-AKT axis by targeting P-REX2a.
miR-338-3p suppresses gastric cancer progression through a PTEN-AKT axis by targeting P-REX2a.
Molecular cancer research : MCR (2014-01-01)
Bo Guo, Liying Liu, Jiayi Yao, Ruili Ma, Dongmin Chang, Zongfang Li, Tusheng Song, Chen Huang
摘要
Results from recent studies suggest that aberrant microRNA expression is common in numerous cancers. Although miR-338-3p has been implicated in hepatocellular carcinoma, its role in gastric cancer is unknown. To this end, we report that miR-338-3p is downregulated in both gastric cancer tissue and cell lines. Forced expression of miR-338-3p inhibited cell proliferation and clonogenicity and induced a G1-S arrest as well as apoptosis in gastric cancer cells. Furthermore, P-Rex2a (PREX2) was identified as a direct target of miR-338-3p, and silencing P-Rex2a resulted in the same biologic effects of miR-338-3p expression in gastric cancer cells. Furthermore, both enforced expression of miR-338-3p or silencing of P-Rex2a resulted in activation of PTEN, leading to a decline in AKT phosphorylation. Also, miR-338-3p markedly inhibited the in vivo tumorigenicity in a nude mouse xenograft model system. These results demonstrate that miR-338-3p affects gastric cancer progression through PTEN-AKT signaling by targeting P-Rex2a in gastric cancer cells, which posits miR-338-3p as a novel strategy for gastric cancer treatment. miR-338-3p acts as a novel tumor suppressor that blocks the growth of gastric cancer cells through PTEN-PI3K signaling by targeting P-Rex2a.