跳轉至內容
Merck
  • Lipoic acid synthetase deficiency causes neonatal-onset epilepsy, defective mitochondrial energy metabolism, and glycine elevation.
Lipoic acid synthetase deficiency causes neonatal-onset epilepsy, defective mitochondrial energy metabolism, and glycine elevation.
American journal of human genetics (2011-12-14)
Johannes A Mayr, Franz A Zimmermann, Christine Fauth, Christa Bergheim, David Meierhofer, Doris Radmayr, Johannes Zschocke, Johannes Koch, Wolfgang Sperl
摘要
Lipoic acid is an essential prosthetic group of four mitochondrial enzymes involved in the oxidative decarboxylation of pyruvate, α-ketoglutarate, and branched chain amino acids and in the glycine cleavage. Lipoic acid is synthesized stepwise within mitochondria through a process that includes lipoic acid synthetase. We identified the homozygous mutation c.746G>A (p.Arg249His) in LIAS in an individual with neonatal-onset epilepsy, muscular hypotonia, lactic acidosis, and elevated glycine concentration in plasma and urine. Investigation of the mitochondrial energy metabolism showed reduced oxidation of pyruvate and decreased pyruvate dehydrogenase complex activity. A pronounced reduction of the prosthetic group lipoamide was found in lipoylated proteins.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
抗硫辛酸兔pAb, liquid, Calbiochem®