跳轉至內容
Merck
  • Half-joule output optical-parametric oscillation by using 10-mm-thick periodically poled Mg-doped congruent LiNbO3.
Half-joule output optical-parametric oscillation by using 10-mm-thick periodically poled Mg-doped congruent LiNbO3.
Optics express (2012-10-06)
Hideki Ishizuki, Takunori Taira
摘要
We present a next generation of large-aperture periodically poled Mg-doped LiNbO3 (PPMgLN) device with 10-mm thickness. Efficient optical parametric oscillation with 540 mJ output energy at 709 mJ pumping by 1.064 µm laser in 10 nanoseconds operation could be demonstrated using the 10-mm-thick PPMgLN with an inversion period of 32.2 µm at total conversion efficiency > 76%. We also confirmed that degradation effect of conversion-efficiency distribution by wedged-inversion structures, which is inevitable in current poling condition of the large-aperture PPMgLN, can be ignored in high-intensity operation.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
铌酸锂, 99.9% trace metals basis