跳轉至內容
Merck
  • Isolation, structure elucidation, and cytotoxic evaluation of furanonaphthoquinones from in vitro plantlets and cultures of Streptocarpus dunnii.
Isolation, structure elucidation, and cytotoxic evaluation of furanonaphthoquinones from in vitro plantlets and cultures of Streptocarpus dunnii.
Journal of natural products (2010-12-23)
Helen Sheridan, Cora Nestor, Lorraine O'Driscoll, Ingrid Hook
摘要
Two new furanonaphthoquinones, (3R)-7-methoxy-α-dunnione (5) and (3R)-6-hydroxy-7-methoxy-α-dunnione (6), along with the known (3R)-dunnione (1), (3R)-α-dunnione (2), (3R)-7-hydroxy-α-dunnione (3), and 1-hydroxy-2-methylanthraquinone (4), were isolated from in vitro cultures of Streptocarpus dunnii. The structures of compounds 5 and 6 were established by spectroscopic means. This is the first report of hydroxylated furanonaphthoquinones in a Streptocarpus species. Compounds 1-3 demonstrated cytotoxic activity against a range of breast cancer and pancreatic tumor cell lines.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
2-甲基蒽醌, technical, ≥95% (HPLC)