跳轉至內容
Merck
Enzymatic determination of sodium and chloride in sweat.
Clinical biochemistry (1996-02-01)
R P Taylor, T J James
摘要
To develop methods based on enzyme activation for the analysis of sweat sodium and chloride using beta-galactosidase and alpha-amylase, respectively. Both were monitored kinetically on the Cobas Fara centrifugal analyzer. The sweat, collected with the Macroduct system, was diluted no more than five-fold for the volumes obtained of 16 to 80 mu L, median 32.5 mu L. The sodium assay utilized a sodium-binding cryptand to maximize linearity. Between-run coefficients of variation (%) at 10, 20, and 50 mmol/L were 3.6, 4.5, and 1.3 for sodium and 7.1, 6.1, and 6.0 for chloride, respectively. The sodium method showed excellent agreement with flame photometry (y = 0.997x + 0.742; r = 0.998), and chloride with a mercuric thiocyanate method (y = 0.995x + 0.485; r = 0.996), giving equivalent discrimination between patients with and without cystic fibrosis. The methods enable the rapid analysis on the same analyzer of both sodium and chloride in a single dilution of sweat collections of low volume.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
4,7,13,16,21-五氧杂-1,10-二氮杂二环[8.8.5]廿三烷, 98%
暫時無法取得訂價和供貨情況