跳轉至內容
Merck
  • Biscatecholate-monohydroxamate mixed ligand siderophore-carbacephalosporin conjugates are selective sideromycin antibiotics that target Acinetobacter baumannii.
Biscatecholate-monohydroxamate mixed ligand siderophore-carbacephalosporin conjugates are selective sideromycin antibiotics that target Acinetobacter baumannii.
Journal of medicinal chemistry (2013-04-26)
Timothy A Wencewicz, Marvin J Miller
摘要
Chemical syntheses and biological evaluation of biscatecholate-monohydroxamate mixed ligand sideromycins utilizing the carbacephalosporin β-lactam antibiotic loracarbef and the fluoroquinolone antibiotic ciprofloxacin are described. The mixed ligand β-lactam sideromycin (1b) had remarkably selective and extremely potent antibacterial activity against the Gram-negative pathogen Acinetobacter baumannii ATCC 17961 (MIC = 0.0078 μM). The antibacterial activity of the β-lactam sideromycin was inversely related to the iron(III) concentration in the testing media and was antagonized by the presence of the competing parent siderophore. These data suggested that active transport of the mixed ligand β-lactam sideromycin across the outer cell membrane of A. baumannii via siderophore-uptake pathways was responsible for the selective and potent antibacterial activity.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
琼脂, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
琼脂, suitable for plant cell culture, powder
Millipore
琼脂, powder, suitable for microbiology
Sigma-Aldrich
诺布尔琼脂
Sigma-Aldrich
琼脂, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
细菌琼脂, for molecular biology
Millipore
琼脂, suitable for microbiology
Sigma-Aldrich
琼脂, ash 2.0-4.5%
Sigma-Aldrich
琼脂, High gel strength, suitable for plant cell culture
Sigma-Aldrich
琼脂, Type A, suitable for plant cell culture
Sigma-Aldrich
选择性琼脂
Millipore
琼脂, suitable for microbiology
Sigma-Aldrich
琼脂, Type M, suitable for plant cell culture
Sigma-Aldrich
琼脂, suitable for plant cell culture
Millipore
琼脂, high purity, low ionic content, low gel strenght, suitable for microbiology