跳轉至內容
Merck
  • A new class of room-temperature multiferroic thin films with bismuth-based supercell structure.
A new class of room-temperature multiferroic thin films with bismuth-based supercell structure.
Advanced materials (Deerfield Beach, Fla.) (2012-11-28)
Aiping Chen, Honghui Zhou, Zhenxing Bi, Yuanyuan Zhu, Zhiping Luo, Adrian Bayraktaroglu, Jamie Phillips, Eun-Mi Choi, Judith L Macmanus-Driscoll, Stephen J Pennycook, Jagdish Narayan, Quanxi Jia, Xinghang Zhang, Haiyan Wang
摘要
Intergrowth of two partially miscible phases of BiFeO(3) and BiMnO(3) gives a new class of room-temperature multiferroic phase, Bi(3) Fe(2) Mn(2) O(10+δ) , which has a unique supercell (SC) structure. The SC heterostructures exhibit simultaneously room-temperature ferrimagnetism and remanent polarization. These results open up a new avenue for exploring room-temperature single-phase multiferroic thin films by controlling the phase mixing of two perovskite BiRO(3) (R = Cr, Mn, Fe, Co, Ni) materials.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
钛酸钙, nanopowder, <100 nm particle size (BET), 99% trace metals basis