跳轉至內容
Merck
  • The application of single-crystal-to-single-crystal transformation towards adjustable SMM properties.
The application of single-crystal-to-single-crystal transformation towards adjustable SMM properties.
Chemical communications (Cambridge, England) (2011-12-14)
Yu-mei Song, Feng Luo, Ming-biao Luo, Zhen-wei Liao, Gong-ming Sun, Xiao-zhao Tian, Yan Zhu, Zi-Jun Yuan, Shu-juan Liu, Wen-yuan Xu, Xue-feng Feng
摘要
Reversible single-crystal-to-single-crystal transformation (SCSC) was for the first time observed between 4f-based molecular magnets.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
1-萘甲酸, 96%