跳轉至內容
Merck
Stapling of a 3(10)-helix with click chemistry.
The Journal of organic chemistry (2011-02-01)
Øyvind Jacobsen, Hiroaki Maekawa, Nien-Hui Ge, Carl Henrik Görbitz, Pål Rongved, Ole Petter Ottersen, Mahmood Amiry-Moghaddam, Jo Klaveness
摘要
Short peptides are important as lead compounds and molecular probes in drug discovery and chemical biology, but their well-known drawbacks, such as high conformational flexibility, protease lability, poor bioavailability and short half-lives in vivo, have prevented their potential from being fully realized. Side chain-to-side chain cyclization, e.g., by ring-closing olefin metathesis, known as stapling, is one approach to increase the biological activity of short peptides that has shown promise when applied to 3(10)- and α-helical peptides. However, atomic resolution structural information on the effect of side chain-to-side chain cyclization in 3(10)-helical peptides is scarce, and reported data suggest that there is significant potential for improvement of existing methodologies. Here, we report a novel stapling methodology for 3(10)-helical peptides using the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) reaction in a model aminoisobutyric acid (Aib) rich peptide and examine the structural effect of side chain-to-side chain cyclization by NMR, X-ray diffraction, linear IR and femtosecond 2D IR spectroscopy. Our data show that the resulting cyclic peptide represents a more ideal 3(10)-helix than its acyclic precursor and other stapled 3(10)-helical peptides reported to date. Side chain-to-side chain stapling by CuAAC should prove useful when applied to 3(10)-helical peptides and protein segments of interest in biomedicine.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
2-氨基异丁酸, 98%