跳轉至內容
Merck
Synthesis of nucleoside-based antiviral drugs in ionic liquids.
Bioorganic & medicinal chemistry letters (2008-09-18)
Vineet Kumar, Sanjay V Malhotra
摘要
Nucleoside-based antiviral drugs have been synthesized using imidazolium-based ionic liquids as reaction medium. The ionic liquids were proved to be better solvents for all the nucleoside in terms of solubility and reaction medium as compared to conventional molecular solvents.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
(E)-5-(2-Bromovinyl)-2′-deoxyuridine