跳轉至內容
Merck
  • Analysis and comparison of active constituents in commercial standardized silymarin extracts by liquid chromatography-electrospray ionization mass spectrometry.
Analysis and comparison of active constituents in commercial standardized silymarin extracts by liquid chromatography-electrospray ionization mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-09-01)
James I Lee, Mahesh Narayan, Jeffrey S Barrett
摘要
A sensitive method for the simultaneous quantitation of six active constituents in commercial silymarin standardized extracts was developed based on liquid chromatography (LC) in combination with mass spectrometry (MS). The six main active constituents, namely, silydianin, silychristin, diastereoisomers of silybin (silybin A and B), and diastereoisomers of isosilybin (isosilybin A and B) were completely separated and quantified by LC/MS. Silymarin obtained from Sigma-Aldrich Co. was evaluated and used as standard reference material for the six individual constituents in comparing the relative content of silymarin and the relative ratio of each constituent in commercial standardized silymarin extracts, respectively. Significant variation was found between different commercial silymarin sources. As a result, this method has proven useful in evaluating and quantifying the six active constituents in commercial milk thistle extracts. The calibration curves were over the range from 0.25 to 100 microg/mL for silychristin and silydianin, and from 0.10 to 100 microg/mL for silybin A, silybin B, isosilybin A and isosilybin B, respectively (r(2)> or =0.9958). For all six active constituents, the overall intra-day precision values, based on the relative standard deviation replicate for four QC levels, ranged from 1.18% to 12.4% and accuracy ranged from 89.4% to 112%. This methodology could easily be incorporated into standardized testing to assess content uniformity including lot-to-lot variation as part of routine process controls as well as a means to describe cross-product variation among the exiting marketed formulations.