跳轉至內容
Merck
  • DNA biosensor for detection of Helicobacter pylori using phen-dione as the electrochemically active ligand in osmium complexes.
DNA biosensor for detection of Helicobacter pylori using phen-dione as the electrochemically active ligand in osmium complexes.
Analytical chemistry (2005-04-15)
M V del Pozo, C Alonso, F Pariente, E Lorenzo
摘要
A surface-based method for the study of the interactions of DNA with redox-active 1,10-phenantroline-5,6-dione (phen-dione) osmium complexes is described. The study was carried out using gold electrodes modified with DNA via adsorption and [Os(bpy)(2)(phe-dione)](3+/2+) (bpy = 2,2'-bipyridyl) or [Os(phen)(2)(phen-dione)](3+/2+) (phen = 1,10-phenantroline) as electrochemical reported molecules. The method, which is simple and reagent-saving, allows the accumulation of osmium complexes within the DNA layer. The amount of osmium complex bound by the adsorbed layer of DNA was determined from the voltammetric charge associated with the osmium redox process of the immobilized metal complex. The quinone moiety of the phen-dione ligand was useful as an indicator for electrochemical DNA sensing because of its redox response at low potentials. A thiol-linked single-stranded Helicobacter pylori DNA probe was immobilized, through S-Au bonds on to a gold electrode (density of modification 86 pmol/cm(2)). Following hybridization with the complementary DNA sequence, the osmium complex was electrochemically accumulated within the double-stranded DNA layer. Electrochemical detection was performed by differential pulse voltammetry over the potential range where the quinone moiety was redox active (i.e., at very low potentials, -0.020 V vs SSCE); with this approach, a sequence of the H. pylori could be quantified over the range from 5 to 20 pmol with a linear correlation of r = 0.9888 and a detection limit of approximately 6 pmol.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
1,10-菲咯啉-5,6-二酮, 97%