跳轉至內容
Merck
  • Apoptosis induced in vivo by photodynamic therapy in normal brain and intracranial tumour tissue.
Apoptosis induced in vivo by photodynamic therapy in normal brain and intracranial tumour tissue.
British journal of cancer (2000-09-20)
L Lilge, M Portnoy, B C Wilson
摘要
The apoptotic response of normal brain and intracranial VX2 tumour following photodynamic therapy (PDT) mediated by 5 different photosensitizers (Photofrin, 5-aminolaevulinic acid (ALA)-induced protoporphyrin IX (PpIX), chloroaluminium phthalocyanine (AlCIPc), Tin Ethyl Etiopurpurin (SnET(2)), and meta -tetra(hydroxyphenyl)chlorin (m THPC)) was evaluated following a previous analysis which investigated the necrotic tissue response to PDT at 24 h post treatment. Free DNA ends, produced by internucleosomal DNA cleavage in apoptotic cells, were stained using a TUNEL (terminal deoxynucleotidyl transferase (TdT)-mediated dUTP nick-end labelling) assay. Confocal laser scanning microscopy (CLSM) was used to quantify the local incidence of apoptosis and determine its spatial distribution throughout the brain. The incidence of apoptosis was confirmed by histopathology, which demonstrated cell shrinkage, pyknosis and karyorrhexis. At 24 h post PDT, AlClPc did not cause any detectable apoptosis, while the other photosensitizers produced varying numbers of apoptotic cells near the region of coagulative necrosis. The apoptotic response did not appear to be related to photosensitizer dose. These results suggest that at this time point, a minimal and fairly localized apoptotic effect is produced in brain tissues, the extent of which depends largely on the particular photosensitizer.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
酞菁氯化铝, Dye content ~85 %