跳轉至內容
Merck
  • Identification and apoptotic potential of T-2 toxin metabolites in human cells.
Identification and apoptotic potential of T-2 toxin metabolites in human cells.
Journal of agricultural and food chemistry (2012-05-04)
Maria Weidner, Tanja Welsch, Florian Hübner, Gerald Schwerdt, Michael Gekle, Hans-Ulrich Humpf
摘要
The mycotoxin T-2 toxin, produced by various Fusarium species, is a widespread contaminant of grain and grain products. Knowledge about its toxicity and metabolism in the human body is crucial for any risk assessment as T-2 toxin can be detected in processed and unprocessed food samples. Cell culture studies using cells of human origin represent a potent model system to study the metabolic fate of T-2 toxin as well as the cytotoxicity in vitro. In this study the metabolism of T-2 toxin was analyzed in a cell line derived from human colon carcinoma cells (HT-29) and primary human renal proximal tubule epithelial cells (RPTEC) using high-performance liquid chromatography coupled with Fourier transformation mass spectrometry (HPLC-FTMS). Both cell types metabolized T-2 toxin to a variety of compounds. Furthermore, cell cycle analysis in RPTEC proved the apoptotic effect of T-2 toxin and its metabolites HT-2 toxin and neosolaniol in micromolar concentrations.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
HT-2 Toxin, ≥98% (HPLC)