跳轉至內容
Merck
Phase I study of H5.020CMV.PDGF-beta to treat venous leg ulcer disease.
Molecular therapy : the journal of the American Society of Gene Therapy (2009-07-30)
David J Margolis, Lee M Morris, Maryte Papadopoulos, Linda Weinberg, Jennifer C Filip, Stephanie A Lang, Sachin S Vaikunth, Timothy M Crombleholme
摘要
Venous leg ulcers are a prevalent nonhealing wound of the lower extremity. Although topically applied growth factors successfully improve wound repair in animal studies, similar studies on humans with venous leg ulcers have not been successful. This study was designed to evaluate the acute safety and biologic feasibility of peri-ulcer injection of a replication-incompetent adenoviral construct expressing platelet-derived growth factor-beta (PDGF-beta). In this phase I study, we demonstrate the initial safety, feasibility, and biologic plausibility of using H5.020CMV.PDGF-beta to treat venous leg ulcer disease.