- Cl-IB-MECA [2-Chloro-N6-(3-iodobenzyl) adenosine-5?-N-methylcarboxamide] Reduces Ischemia/Reperfusion Injury in Mice by Activating the A Adenosine Receptor
Cl-IB-MECA [2-Chloro-N6-(3-iodobenzyl) adenosine-5?-N-methylcarboxamide] Reduces Ischemia/Reperfusion Injury in Mice by Activating the A Adenosine Receptor
Journal of Pharmacology and Experimental Therapeutics (2006)
Ge ZD, et al.