跳轉至內容
Merck
  • TGFβ blocks IFNα/β release and tumor rejection in spontaneous mammary tumors.
TGFβ blocks IFNα/β release and tumor rejection in spontaneous mammary tumors.
Nature communications (2019-09-13)
Marion V Guerin, Fabienne Regnier, Vincent Feuillet, Lene Vimeux, Julia M Weiss, Georges Bismuth, Gregoire Altan-Bonnet, Thomas Guilbert, Maxime Thoreau, Veronica Finisguerra, Emmanuel Donnadieu, Alain Trautmann, Nadège Bercovici
摘要
Type I interferons (IFN) are being rediscovered as potent anti-tumoral agents. Activation of the STimulator of INterferon Genes (STING) by DMXAA (5,6-dimethylxanthenone-4-acetic acid) can induce strong production of IFNα/β and rejection of transplanted primary tumors. In the present study, we address whether targeting STING with DMXAA also leads to the regression of spontaneous MMTV-PyMT mammary tumors. We show that these tumors are refractory to DMXAA-induced regression. This is due to a blockade in the phosphorylation of IRF3 and the ensuing IFNα/β production. Mechanistically, we identify TGFβ, which is abundant in spontaneous tumors, as a key molecule limiting this IFN-induced tumor regression by DMXAA. Finally, blocking TGFβ restores the production of IFNα by activated MHCII+ tumor-associated macrophages, and enables tumor regression induced by STING activation. On the basis of these findings, we propose that type I IFN-dependent cancer therapies could be greatly improved by combinations including the blockade of TGFβ.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
DMXAA, ≥98% (HPLC), solid