跳轉至內容
Merck
  • Conformation-dependent cleavage of staphylococcal nuclease with a disulfide-linked iron chelate.
Conformation-dependent cleavage of staphylococcal nuclease with a disulfide-linked iron chelate.
Proceedings of the National Academy of Sciences of the United States of America (1992-07-15)
M R Ermácora, J M Delfino, B Cuenoud, A Schepartz, R O Fox
摘要
We report the synthesis and evaluation of (EDTA-2-aminoethyl) 2-pyridyl disulfide. By using this easily prepared cysteine-specific hydrophilic reagent, an ethylenediaminetriacetic acid-Fe3+ complex (EDTA-Fe) was covalently attached to a single genetically engineered cysteine residue in staphylococcal nuclease. Upon addition of the iron reductant ascorbate, the nuclease-EDTA-Fe conjugate underwent a protein self-cleavage reaction mediated by reactive oxygen species. Sequence analysis of the products indicated that cleavage occurs close in tertiary structure to the EDTA-Fe attachment site. In the presence of denaturants, the cleavage pattern changes and the reaction is limited to residues proximal in sequence to the cysteine attachment site. These results indicate that intramolecular protein cleavage reactions mediated by EDTA-Fe can be used to evaluate changes in protein conformation. The reagent described should be a useful tool in the structural mapping of nonnative protein states populated at equilibrium, such as the molten globule, that are frequently refractory to conventional structure analysis.