跳轉至內容
Merck
  • Nickel-catalyzed C-O activation of phenol derivatives with potassium heteroaryltrifluoroborates.
Nickel-catalyzed C-O activation of phenol derivatives with potassium heteroaryltrifluoroborates.
Organic letters (2010-08-19)
Gary A Molander, Floriane Beaumard
摘要
A general method based on nickel-catalyzed C-O activation of various phenol derivatives with potassium (hetero)aryltrifluoroborates has been developed. A large number of heterobiaryls can be easily obtained with yields up to 99% using methanesulfonate cross-coupling partners.