Merck
  • JNK inhibitor alleviates apoptosis of fetal neural stem cells induced by emulsified isoflurane.
JNK inhibitor alleviates apoptosis of fetal neural stem cells induced by emulsified isoflurane.
Oncotarget (2017-12-08)
Lei Zhou, Zeyong Yang, Xianfu Lu, Xingxing Li, Xiaohu An, Jing Chai, Qiling Yang, Shikai Yan, Yuanhai Li
ABSTRACT
Isoflurane can provide both neuroprotection and neurotoxicity in various culture models and in rodent developing brains. Emulsified Isoflurane (EI) is an emulsion formulation of isoflurane, while its underlying molecular mechanism of developemental nerve toxicity largely remains unclear. We hypothesized that EI induced fetal neural stem cells (FNSCs) apoptosis, endoplasmic reticulum (ER) stress and c-Jun N-terminal kinase (JNK) activation. FNSCs were isolated from the cortex of SD rats during 14 days of gestation. The cell viability, cell apoptotic rates and the expression of apoptosis-related protein Caspase3, inositol requiring enzyme 1 (IRE1), poly (adenosine diphosphate-ribose) polymerase (PARP), Bax, Bcl-2, JNK, p-JNK and XBP1 were determined. Specific inhibition was performed by siRNA-targeting of JNK in FNSCs. EI could increase the p-JNK, JNK and caspase3 protein expression, the JNK pathway was activated by EI, and EI-induced apoptosis was blocked by inhibiting JNK pathway with SP600125 or JNK-small interfering RNA (siRNA), EI enhanced the level of IRE1, PARP, Bax/Bcl-2 and XBP1, which led FNSCs to apoptosis and ER stress. Meanwhile, dilatation of the ER lumens in FNSCs treated by EI for 24 h was significant. G 1. Prolonged treatment with high-dose EI decreased the survival level of FNSCs by inducing apoptosis and inhibiting proliferation via the JNK signaling pathway. 2. EI induced ER stress and sequent JNK activation. 3. JNK inhibiting might be applied for improving therapeutic outcomes in anesthestics-induced neurotoxicity.