Merck
  • Genetic and pharmacological inhibition of microRNA-92a maintains podocyte cell cycle quiescence and limits crescentic glomerulonephritis.
Genetic and pharmacological inhibition of microRNA-92a maintains podocyte cell cycle quiescence and limits crescentic glomerulonephritis.
Nature communications (2017-12-01)
Carole Henique, Guillaume Bollée, Xavier Loyer, Florian Grahammer, Neeraj Dhaun, Marine Camus, Julien Vernerey, Léa Guyonnet, François Gaillard, Hélène Lazareth, Charlotte Meyer, Imane Bensaada, Luc Legrès, Takashi Satoh, Shizuo Akira, Patrick Bruneval, Stefanie Dimmeler, Alain Tedgui, Alexandre Karras, Eric Thervet, Dominique Nochy, Tobias B Huber, Laurent Mesnard, Olivia Lenoir, Pierre-Louis Tharaux
ABSTRACT
Crescentic rapidly progressive glomerulonephritis (RPGN) represents the most aggressive form of acquired glomerular disease. While most therapeutic approaches involve potentially toxic immunosuppressive strategies, the pathophysiology remains incompletely understood. Podocytes are glomerular epithelial cells that are normally growth-arrested because of the expression of cyclin-dependent kinase (CDK) inhibitors. An exception is in RPGN where podocytes undergo a deregulation of their differentiated phenotype and proliferate. Here we demonstrate that microRNA-92a (miR-92a) is enriched in podocytes of patients and mice with RPGN. The CDK inhibitor p57
Sigma-Aldrich
MISSION® esiRNA, targeting human CORO1A