Merck
  • Comparative evaluation of T11 target structure and its deglycosylated derivative nullifies the importance of glycan moieties in immunotherapeutic efficacy.
Comparative evaluation of T11 target structure and its deglycosylated derivative nullifies the importance of glycan moieties in immunotherapeutic efficacy.
Acta biochimica et biophysica Sinica (2012-01-20)
Sirshendu Chatterjee, Sagar Acharya, Pankaj Kumar, Ananya Chatterjee, Suhnrita Chaudhuri, Anirban Ghosh, Swapna Chaudhuri
ABSTRACT
Sheep red blood cell (SRBC), a non-specific biological response modifier that has long been used as a classical antigen, has been shown to exert an immunomodulatory and anti-tumor activities in experimental animals. The active component of SRBC, which is responsible for such effects, was found to be a cell surface acidic glycoprotein molecule, known as T11 target structure (T11TS). In the present study, T11TS was isolated and purified to homogeneity using a five-step protocol involving isolation of sheep erythrocyte membrane from packed cell volume, 20% ammonium sulfate cut of the crude membrane proteins mixture, immunoaffinity purification using mouse anti-sheep CD58 mAb (L180/1) tagged matrix, preparative gel electrophoresis, and gel electroelution process. Finally, the purity and identity of the proteins were confirmed by the matrix-assisted laser desorption/ionization (MALDI) mass spectrometric analysis. The in silico glycosylation site analysis showed that the extracellular domain contained three N-glycosylation sites (N-12, N-62, and N-111) and one O-glycosylation site (T-107). However, the experimental analysis negated the presence of O-linked glycan moieties on T11TS. To investigate the role of glycan moieties in the current immunotherapeutic regime, T11TS and its deglycosylated form (dT11TS) were administered intraperitoneally (i.p.) in N-ethyl-N-nitrosourea-induced immune-compromised mice at 0.4 mg/kg body weight. It was observed that both the forms of T11TS could activate the compromised immune status of mice by augmenting immune receptor expression (CD2, CD25, CD8, and CD11b), T-helper 1 shift of cytokine network, enhanced cytotoxicity, and phagocytosis activity. Therefore, the results nullify the active involvement of the N-linked glycan moieties in immunotherapeutic efficacy of T11TS.
Product Description
G-CAPSULE electroelution device