Merck
  • Multiparametric assessment of the anti-glioma properties of OKN007 by magnetic resonance imaging.
Multiparametric assessment of the anti-glioma properties of OKN007 by magnetic resonance imaging.
Journal of magnetic resonance imaging : JMRI (2010-04-08)
Philippe Garteiser, Sabrina Doblas, Yasuko Watanabe, Debbie Saunders, Jessica Hoyle, Megan Lerner, Ting He, Robert A Floyd, Rheal A Towner
ABSTRACT
To demonstrate that OKN007, a disulfonyl derivative of phenyl-tert-butyl nitrone (PBN), has anti-glioma activity in the clinically relevant C6 rat glioma model using multi-parametric magnetic resonance imaging. Twenty-one rats were intracerebrally implanted with C6 cells and administered OKN007 or kept as controls. Animals were monitored with MRI at 7 Tesla (T), using morphologic, diffusion-weighted and perfusion imaging, followed by histology and Western blots of angiogenesis and inflammatory markers. OKN007 was found to decrease tumor volumes and increase survival. The glioma tissues of OKN007-treated rats were found to have longitudinal apparent diffusion coefficients (ADC(z)) of 0.76 +/- 0.06 x 10(-3) mm(2)/s, similar to the contralateral tissue and significantly smaller than untreated gliomas (0.97 +/- 0.13 x 10(-3) mm(2)/s). They had higher perfusion rates (66 +/- 4 mL/100 g.min) than untreated gliomas (26 +/- 7 mL/100 g.min). All examined molecular markers were decreased in OKN007-treated rat gliomas, compared with elevated levels in untreated rats. MRI assessment was successfully used to monitor a decrease in tumor growth, and corresponding alterations in ADC and perfusion rates in rat C6 gliomas treated with the anti-glioma agent, OKN007.
Sigma-Aldrich
HPN-07, ≥98% (HPLC)