Merck
Reversion of antibiotic resistance in
Science (New York, N.Y.) (2017-03-18)
Nicolas Blondiaux, Martin Moune, Matthieu Desroses, Rosangela Frita, Marion Flipo, Vanessa Mathys, Karine Soetaert, Mehdi Kiass, Vincent Delorme, Kamel Djaout, Vincent Trebosc, Christian Kemmer, René Wintjens, Alexandre Wohlkönig, Rudy Antoine, Ludovic Huot, David Hot, Mireia Coscolla, Julia Feldmann, Sebastien Gagneux, Camille Locht, Priscille Brodin, Marc Gitzinger, Benoit Déprez, Nicolas Willand, Alain R Baulard
ABSTRACT
Antibiotic resistance is one of the biggest threats to human health globally. Alarmingly, multidrug-resistant and extensively drug-resistant