Merck
Pd(II)-Catalyzed Enantioselective C(sp
Journal of the American Chemical Society (2017-02-18)
Jian He, Qian Shao, Qingfeng Wu, Jin-Quan Yu
ABSTRACT
Pd(II)-catalyzed enantioselective borylation of C(sp