Merck
  • Dihydroorotate dehydrogenase Inhibitors Target c-Myc and Arrest Melanoma, Myeloma and Lymphoma cells at S-phase.
Dihydroorotate dehydrogenase Inhibitors Target c-Myc and Arrest Melanoma, Myeloma and Lymphoma cells at S-phase.
Journal of Cancer (2017-09-21)
Mathura Subangari Dorasamy, Bhavesh Choudhary, Kavitha Nellore, Hosahalli Subramanya, Pooi-Fong Wong
ABSTRACT
Dihydroorotate dehydrogenase (DHODH) is a rate-limiting enzyme in the
Sigma-Aldrich
Triton X-100, laboratory grade