Merck
  • Construction and evaluation of pH-sensitive immunoliposomes for enhanced delivery of anticancer drug to ErbB2 over-expressing breast cancer cells.
Construction and evaluation of pH-sensitive immunoliposomes for enhanced delivery of anticancer drug to ErbB2 over-expressing breast cancer cells.
Nanomedicine : nanotechnology, biology, and medicine (2016-12-15)
Tianshu Li, Takuya Amari, Kentaro Semba, Tadashi Yamamoto, Shinji Takeoka
ABSTRACT
1,5-Dihexadecyl N,N-diglutamyl-lysyl-L-glutamate (GGLG) liposomes were previously developed to enhance drug delivery efficiency in tumor cells owing to its pH-responsive properties. Herein, we report the modification of GGLG liposomes by conjugating a Fab' fragment of an ErbB2 antibody to the terminus of PEG (polyethylene glycol)-lipid (Fab'-GGLG liposomes). The conjugation of Fab' fragments did not affect the antibody activity, drug (doxorubicin, DOX) encapsulation efficiency, stability during storage or pH-sensitivity. However, the binding affinity of Fab'-GGLG liposomes was enhanced to ErbB2-overexpressing HCC1954 cells specifically, and the cell association increased 10-fold in comparison to GGLG liposomes. Consequently, intracellular DOX delivery was enhanced, with an increased cytotoxicity in HCC1954 cells (i.e., IC
Sigma-Aldrich
Maleimide, 99%