Merck
Neuroprotective effect of G
Neurological research (2017-07-20)
Guangsheng Gao, Huaihai Fan, Xiaoying Zhang, Fusen Zhang, Haiyan Wu, Feng Qi, Lei Zhao, Yun Li
ABSTRACT
Humanin (HN) has been identified to suppress neuron death. Gly Rats were randomly divided into 13 groups: Sham group, GI groups and HNG groups. Both GI group and HNG groups included six time points (1, 3, 6, 12, 24, and 72 h). At 24 h after reperfusion, Nissl staining was used to observe positive neurons, and p-STAT3, MCL-1, SOCS3, Bax and Caspase-3 in different groups were detected by immunohistochemistry. qRT-PCR and western blot were used to evaluate the expression of STAT3, p-STAT3, MCL-1, and SOCS3. The immunohistochemistry also showed a significant increase in Bax (0.29 ± 0.007 vs. 0.22 ± 0.007, P < 0.01) and Caspase-3 (0.24 ± 0.02 vs. 0.18 ± 0.006, P < 0.01) in GI group compared with Sham group, while Bax (0.26 ± 0.01 vs. 0.29 ± 0.008, P < 0.01) and Caspase-3 (0.20 ± 0.008 vs. 0.24 ± 0.02, P < 0.01) were significantly decreased by HNG-treatment compared with GI group. Along with immunohistochemistry, western blot and qRT-PCR indicated that the protein and mRNA levels of STAT3, MCL-1, and SOCS3 were up-regulated after administration of HNG at six time points after global cerebral I/R in rat. HNG might exert neuroprotective effects through alleviating apoptosis and activating of SOCS3 - STAT3 - MCL-1 signal transduction pathway. Highlights (1) Cerebral ischemia led to neuronal loss in hippocampal CA1 region of rats. (2) HNG had neuroprotective effects on ischemia/reperfusion rats. (3) The protective effect of HNG might be related to the SOCS3 - STAT3 - MCL-1 pathway.
Sigma-Aldrich
[Gly14]-Humanin G human, ≥95% (HPLC), powder