Merck
  • Rescue of impaired sociability and anxiety-like behavior in adult cacna1c-deficient mice by pharmacologically targeting eIF2α.
Rescue of impaired sociability and anxiety-like behavior in adult cacna1c-deficient mice by pharmacologically targeting eIF2α.
Molecular psychiatry (2017-06-07)
Z D Kabir, A Che, D K Fischer, R C Rice, B K Rizzo, M Byrne, M J Glass, N V De Marco Garcia, A M Rajadhyaksha
ABSTRACT
CACNA1C, encoding the Ca
Sigma-Aldrich
ISRIB, ≥98% (HPLC)