Merck
  • Inhibitory effect of 4-(2-aminoethyl)-benzenesulfonyl fluoride, a serine protease inhibitor, on PI3K inhibitor-induced CHOP expression.
Inhibitory effect of 4-(2-aminoethyl)-benzenesulfonyl fluoride, a serine protease inhibitor, on PI3K inhibitor-induced CHOP expression.
European journal of pharmacology (2006-11-18)
Toru Hosoi, Kanae Hyoda, Yasunobu Okuma, Yasuyuki Nomura, Koichiro Ozawa
ABSTRACT
Endoplasmic reticulum stress contributes to several diseases such as neurodegenerative disorders and diabetes. In the previous report, we found that phosphatidylinositol 3-kinase (PI3K) down-regulation is important for inducing CHOP expression, an endoplasmic reticulum stress-induced transcription factor. In the present study, we investigated the effect of 4-(2-aminoethyl)-benzenesulfonyl fluoride (AEBSF), a serine protease inhibitor, on PI3K inhibitor-induced CHOP expression. We found that AEBSF completely inhibited PI3K inhibitor-induced CHOP expression at both mRNA and protein levels. It is suggested that AEBSF is an important drug from a pharmacological point of view and the results may have important implications for understanding endoplasmic reticulum stress-related diseases.