Merck
  • A highly sensitive colorimetric determination of serum zinc using water-soluble pyridylazo dye.
A highly sensitive colorimetric determination of serum zinc using water-soluble pyridylazo dye.
Clinica chimica acta; international journal of clinical chemistry (1982-03-26)
T Makino, M Saito, D Horiguchi, K Kina
ABSTRACT
A colorimetric method for precise and accurate determination of zinc in serum is presented. Only 0.3 ml of sample is necessary, because of the use of a new, highly sensitive reagent, 2-(5-bromo-2-pyridylazo)-5-(N-n-propyl-N-3-sulfopropylamino)-phenol (epsilon 554nm = 1.3 X 10(5) 1 . mol-1 . cm-1), which is water-soluble and has long-term stability. Interference of iron and copper in serum can be removed by co-precipitation of the iron fluoride complex with trichloroacetic acid precipitated proteins and the copper dithiocarboxy sarcosine complex, respectively. Within-run and day-to-day precision (CV) are in the range of 0.3-3.5% and 1.9-3.1%, respectively, depending on the serum zinc content. A good correlation (r = 0.98, p less than 0.05) was obtained between this method and atomic absorption spectrometry. In contrast to previous colorimetric methods, the present method does not involve heating, extraction with organic solvents, or a cyanide masking system.
Product Description
Supelco
2-(5-Bromo-2-pyridylazo)-5-[N-propyl-N-(3-sulfopropyl)amino]phenol disodium salt dihydrate, for spectrophotometric det. of Zn(II), Cu(II), Fe(II), Co(II), H2O2, ≥95.0% (HPLC)