Merck
  • Determination of 5-aminoimidazole-4-carboxamide in human plasma by ion-pair extraction and LC-MS/MS.
Determination of 5-aminoimidazole-4-carboxamide in human plasma by ion-pair extraction and LC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2007-06-15)
Xijing Chen, Jing Jing, Weichao Ren, De-En Han, Ning Jing, Guangji Wang
ABSTRACT
A liquid chromatography/tandem mass spectrometry (LC-MS/MS) method was established for the determination of 5-aminoimidazole-4-carboxamide (AICA) in human plasma. The method included a solvent extraction of AICA as an ion pair with 1-pentanesulfonate ion and a separation on a Hypersil ODS2 column with the mobile phase of methanol-water (68:32, v/v). Determination was performed using an electrospray ionization source in positive ion mode (ESI(+)). Multiple reaction monitoring (MRM) was utilized for the detection monitoring m/z at 127-->110 for AICA, and 172-->128 for IS. The calibration curve was linear within a range from 20 to 2000 ng/mL and the limit of quantity for AICA in plasma was 20 ng/mL. RSD of intra-assay and inter-assay were no more than 5.90% and 5.65%.
Product Description
Supelco
Sodium 1-pentanesulfonate monohydrate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)